Please login to your online account to display your discounted pricing

anti-TIM-1/KIM-1/HAVCR, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056421SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056421T

  • / Each
 

Description & Specifications

Specifications

Immunogen A synthetic peptide (SPLYSYTTDGNDTVT) of human TIM-1 protein was used as immunogen for this antibody. This sequence corresponds to amino acids (aa) 248-262 of NP_036338 and aa 84-98 of EAW61615.1. Isoforms of TIM-1 have been described. For example, NP_036338 is 364 aa and EAW61615.1 is 194 aa. EAW61615.1 is referred to as isoform CRA_a.
Antigen TIM-1/KIM-1/HAVCR
Format Purified
Formulation PBS containing 0.05% BSA
Host Species Rabbit
Applications Western Blotting
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 26762
Test Specificity The amino acid sequence (SPLYSYTTDGNDTVT) used as immunogen for this antibody is 100% homologous in human, 93% homologous in vervet/green monkey and 80% homologous in chimp. Isoforms of different molecular weights have been described for TIM-1. The sequence used for immunogen is conserved between TIM-1 (364aa, NP_036338) and TIM-1 isoform CRA_a (194aa, EAW61615.1). Therefore, the molecular weight observed in western blots may vary depending on the forms present. Reacts with Human and mouse. Predicted to react with New World Monkey.
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse