Please login to your online account to display your discounted pricing

anti-TIGAR/C12orf5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP149534

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP149534

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Gene ID (Entrez) 57103
Concentration 0.5mg/mL
Primary or Secondary Primary
Format Affinity Purified
Cross Reactivity Human
Cross Reactivity Mouse
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Formulation PBS, 30% glycerol
Gene Accession No. Q9NQ88
Quantity 0.1mL
Regulatory Status RUO
Host Species Rabbit
Antigen TIGAR/C12orf5
Immunogen A genomic peptide made to an internal region of the human TIGAR protein (within residues 50-200). [Swiss-Prot Q9NQ88]