missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TIGAR/C12orf5, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 220-270 of human TIGAR was used as the immunogen.
Antigen TIGAR/C12orf5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224507 Novus Biologicals Canada
NBP224507
0.1mg Each for $255.80
NBP224507SS Novus Biologicals Canada
NBP224507SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
TIGAR/C12orf5
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_b) and chimpanzee, 94% homologous in rat, cow, dog, mouse, and horse, and 88% homologous in opossum.
57103
0.5mg/mL
A portion of amino acids 220-270 of human TIGAR was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
Q9NQ88
100μg in 200μL PBS containing 0.2% gelatin