missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TIGAR/C12orf5, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human TIGAR protein (within residues 50-200). [Swiss-Prot Q9NQ88]
Antigen TIGAR/C12orf5
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP149534 Novus Biologicals Canada
NBP149534
0.1mL Each for $309.05
NBP149534SS Novus Biologicals Canada
NBP149534SS
0.025mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
TIGAR/C12orf5
Affinity Purified
RUO
Q9NQ88
PBS, 30% glycerol
A genomic peptide made to an internal region of the human TIGAR protein (within residues 50-200). [Swiss-Prot Q9NQ88]
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
57103
0.5mg/mL