Please login to your online account to display your discounted pricing

anti-TIGAR/C12orf5, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224507

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224507

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q9NQ88
Primary or Secondary Primary
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_b) and chimpanzee, 94% homologous in rat, cow, dog, mouse, and horse, and 88% homologous in opossum.
Concentration 0.5mg/mL
Gene ID (Entrez) 57103
Conjugate Unlabeled
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Primate
Regulatory Status RUO
Immunogen A portion of amino acids 220-270 of human TIGAR was used as the immunogen.
Host Species Rabbit
Antigen TIGAR/C12orf5
Quantity 0.1mg
Format Affinity Purified