missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Tie-2, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human TIE2 protein (within residues 250-400). [Swiss-Prot Q02763]
Antigen Tie-2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP169753SS Novus Biologicals Canada
NBP169753SS
0.025mL Each for $139.00
NBP169753 Novus Biologicals Canada
NBP169753
0.1mL Each for $329.00
Specifications Description & Specifications

Specifications

Rabbit
Tie-2
Affinity Purified
RUO
Q02763
PBS, 30% glycerol
A genomic peptide made to an internal region of the human TIE2 protein (within residues 250-400). [Swiss-Prot Q02763]
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin),Simple Western,Western Blot
7010
1.09mg/mL