Please login to your online account to display your discounted pricing

anti-thymocyte selection-associated high mobility group box, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 475-525 from human TOX was used as the immunogen for this antibody.
Antigen thymocyte selection-associated high mobility group box
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227156SS

 
novus biologicals canada
NBP227156SS
Unlabeled 0.025mg Each for $109.00

NBP227156

 
novus biologicals canada
NBP227156
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 475-525 from human TOX was used as the immunogen for this antibody.
Antigen thymocyte selection-associated high mobility group box
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The TOX amino acid sequence used as immunogen for this antibody is 100% homologous in human, chimpanzee, gorilla and monkey, 95% in bovine, dog, and horse, and 90% in mouse and rat.
Gene Accession No. O94900
Gene ID (Entrez) 9760
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL