Please login to your online account to display your discounted pricing

anti-thymocyte selection-associated high mobility group box, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227156SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227156SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Antigen thymocyte selection-associated high mobility group box
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Gene Accession No. O94900
Host Species Rabbit
Applications Western Blotting
Immunogen A portion of amino acids 475-525 from human TOX was used as the immunogen for this antibody.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9760
Test Specificity The TOX amino acid sequence used as immunogen for this antibody is 100% homologous in human, chimpanzee, gorilla and monkey, 95% in bovine, dog, and horse, and 90% in mouse and rat.
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human