Please login to your online account to display your discounted pricing

anti-thymocyte selection-associated high mobility group box, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227156

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227156

  • / Each
 

Description & Specifications

Specifications

Antigen thymocyte selection-associated high mobility group box
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Gene Accession No. O94900
Host Species Rabbit
Immunogen A portion of amino acids 475-525 from human TOX was used as the immunogen for this antibody.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 9760
Test Specificity The TOX amino acid sequence used as immunogen for this antibody is 100% homologous in human, chimpanzee, gorilla and monkey, 95% in bovine, dog, and horse, and 90% in mouse and rat.
Target Species Human