Please login to your online account to display your discounted pricing

anti-Thrombomodulin/BDCA-3, Clone: 501733, R&D Systems


Manufacturer: r & d systems  RDSMAB3947100

 View more versions of this product

Catalog No. MAB3947100

  • / Each
 

Description & Specifications

Specifications

Format Purified
Isotype IgG1
Cross Reactivity Human
Host Species Mouse
Conjugate Unlabeled
Regulatory Status RUO
Primary or Secondary Primary
Clone 501733
Immunogen Mouse myeloma cell line NS0-derived recombinant human Thrombomodulin/BDCA-3. Ala19-Ser515
Quantity 100μg
Antigen Thrombomodulin/BDCA-3
Monoclonal or Polyclonal Monoclonal
Purification Method Protein A or G purified from hybridoma culture supernatant

Thrombomodulin/BDCA-3 Monoclonal antibody MAB3947100 Specifically detects Thrombomodulin/BDCA-3 in Human samples. It is validated for ELISA Capture, Flow Cytometry, Immunohistochemistry