Please login to your online account to display your discounted pricing

anti-Thrombomodulin/BDCA-3, Clone: 501733, R&D Systems

$326.67 - $326.67

Specifications

Antigen Thrombomodulin/BDCA-3
Clone 501733
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MAB3947100

 
r & d systems
RDSMAB3947100
Unlabeled 100μg Each for $326.67
Description & Specifications

Specifications

Antigen Thrombomodulin/BDCA-3
Clone 501733
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant human Thrombomodulin/BDCA-3. Ala19-Ser515
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

ELISA Capture, Flow Cytometry, Immunohistochemistry