missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-THEM2, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 100-150 of human THEM2 was used as the immunogen for the antibody.
Antigen THEM2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224655 Novus Biologicals Canada
NBP224655
0.1mg Each for $257.41
NBP224655SS Novus Biologicals Canada
NBP224655SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
THEM2
Purified
RUO
The amino acid sequence used as immunogen for the antibody is 100% homologous in human,mouse, rat and bovine.
55856
0.5mg/mL
A portion of amino acid 100-150 of human THEM2 was used as the immunogen for the antibody.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
Q9NPJ3
100μg antibody in 200μL PBS containing 0.05% BSA