Please login to your online account to display your discounted pricing

anti-THEM2, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 25-75 of human THEM2 was used as the immunogen.
Antigen THEM2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224609

 
novus biologicals canada
NBP224609
Unlabeled 0.1mg Each for $257.41

NBP224609SS

 
novus biologicals canada
NBP224609SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 25-75 of human THEM2 was used as the immunogen.
Antigen THEM2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and 88% homologous in mouse and rat (predicted).
Gene Accession No. Q9NPJ3
Gene ID (Entrez) 55856
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL