Please login to your online account to display your discounted pricing

anti-THEM2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224609SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224609SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q9NPJ3
Antigen THEM2
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Applications Western Blotting
Immunogen A portion of amino acids 25-75 of human THEM2 was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 55856
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and 88% homologous in mouse and rat (predicted).
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat