Please login to your online account to display your discounted pricing

anti-TGR5/GPBAR1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224577SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224577SS

  • / Each
 

Description & Specifications

Specifications

Antigen TGR5/GPBAR1
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. Q8TDU6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 302-366 of human GPR19 was used as the immunogen for this antibody.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 151306
Target Species Human