Please login to your online account to display your discounted pricing

anti-TGR5/GPBAR1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223668SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223668SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Antigen TGR5/GPBAR1
Format Antiserum
Formulation 100μL of crude rabbit sera with 50% glycerol
Gene Accession No. Q8TDU6
Host Species Rabbit
Applications Western Blotting
Immunogen A portion of amino acid 1-50 of human BG37/TGR5 was used as the immunogen.
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151306
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee and 72% homologous in monkey.
Conjugate Unlabeled