Please login to your online account to display your discounted pricing

anti-TGR5/GPBAR1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223662SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223662SS

  • / Each
 

Description & Specifications

Specifications

Format Antiserum
Formulation 100μL of crude rabbit sera with 50% glycerol
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Antigen TGR5/GPBAR1
Gene Accession No. Q8TDU6
Host Species Rabbit
Applications Western Blotting
Immunogen A portion of amino acid 200-250 of human BG37/TGR5 was used as the immunogen.
Quantity 0.025mL
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 151306
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, cat, chicken, rat, mouse, platypus, rhesus monkey and guinea pig, 93% homologous in Xenopus and 86% homologous in zebrafish.