Please login to your online account to display your discounted pricing

anti-TGR5/GPBAR1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223662

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223662

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 151306
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Regulatory Status RUO
Quantity 0.1mL
Gene Accession No. Q8TDU6
Conjugate Unlabeled
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, cat, chicken, rat, mouse, platypus, rhesus monkey and guinea pig, 93% homologous in Xenopus and 86% homologous in zebrafish.
Formulation 100μL of crude rabbit sera with 50% glycerol
Host Species Rabbit
Immunogen A portion of amino acid 200-250 of human BG37/TGR5 was used as the immunogen.
Antigen TGR5/GPBAR1
Format Antiserum
Cross Reactivity Human
Cross Reactivity Mouse
Applications Western Blotting