Please login to your online account to display your discounted pricing

anti-TGR5/GPBAR1, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 302-366 of human GPR19 was used as the immunogen for this antibody.
Antigen TGR5/GPBAR1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224577

 
novus biologicals canada
NBP224577
Unlabeled 0.1mg Each for $307.00

NBP224577SS

 
novus biologicals canada
NBP224577SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 302-366 of human GPR19 was used as the immunogen for this antibody.
Antigen TGR5/GPBAR1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. Q8TDU6
Gene ID (Entrez) 151306
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL