Please login to your online account to display your discounted pricing

anti-TGR5/GPBAR1, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 1-50 of human BG37/TGR5 was used as the immunogen.
Antigen TGR5/GPBAR1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223668

 
novus biologicals canada
NBP223668
Unlabeled 0.1mL Each for $259.02

NBP223668SS

 
novus biologicals canada
NBP223668SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 1-50 of human BG37/TGR5 was used as the immunogen.
Antigen TGR5/GPBAR1
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee and 72% homologous in monkey.
Gene Accession No. Q8TDU6
Gene ID (Entrez) 151306
Formulation 100μL of crude rabbit sera with 50% glycerol