Please login to your online account to display your discounted pricing

anti-TGR5/GPBAR1, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 200-250 of human BG37/TGR5 was used as the immunogen.
Antigen TGR5/GPBAR1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223662

 
novus biologicals canada
NBP223662
Unlabeled 0.1mL Each for $259.02

NBP223662SS

 
novus biologicals canada
NBP223662SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 200-250 of human BG37/TGR5 was used as the immunogen.
Antigen TGR5/GPBAR1
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, cat, chicken, rat, mouse, platypus, rhesus monkey and guinea pig, 93% homologous in Xenopus and 86% homologous in zebrafish.
Gene Accession No. Q8TDU6
Gene ID (Entrez) 151306
Formulation 100μL of crude rabbit sera with 50% glycerol