Please login to your online account to display your discounted pricing

anti-TGN46, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP149643SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP149643SS

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Gene ID (Entrez) 10618
Cross Reactivity Human
Antigen TGN46
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. O43493
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human TGN46 protein (within residues 200-350). [Swiss-Prot O43493]
Quantity 0.025mL
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Concentration 0.66mg/mL
Conjugate Unlabeled