Please login to your online account to display your discounted pricing

anti-TGF-beta RII, Polyclonal, Novus Biologicals™

$171.00 - $171.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide
Antigen TGF-beta RII
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10091994T

 
novus biologicals canada
NB10091994SS
Unlabeled 0.02mg Each for $171.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide
Antigen TGF-beta RII
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. P37173
Gene ID (Entrez) 7048
Formulation PBS, pH 7.2.
Concentration 1.0mg/mL