Please login to your online account to display your discounted pricing

anti-TGF-beta RII, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10091994SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10091994T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7048
Monoclonal or Polyclonal Polyclonal
Gene Accession No. P37173
Quantity 0.02mg
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen TGF-beta RII
Immunogen Synthetic peptide
Conjugate Unlabeled
Format Affinity Purified
Regulatory Status RUO
Host Species Rabbit
Concentration 1.0mg/mL
Primary or Secondary Primary
Formulation PBS, pH 7.2.