Please login to your online account to display your discounted pricing

anti-TGF-beta 2, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAB112NA

 View more versions of this product

Catalog No. AB112NA

  • / Each
 

Description & Specifications

Specifications

Immunogen Porcine platelet-derived TGF-β2
Regulatory Status RUO
Format Purified
Quantity 1mg
Host Species Goat
Antigen TGF-beta 2
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Isotype IgG