missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TGF-beta 2, Polyclonal, R&D Systems

$520.12 - $520.12

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Conjugate Quantity Price Quantity  
AB12NA R & D Systems
RDSAB12NA
Unlabeled 1mg Each for $520.12
AB112NA R & D Systems
RDSAB112NA
Unlabeled 1mg Each for $520.12
Specifications

Description

Host Species: Goat
Immunogen:Porcine platelet-derived TGF-β2

Applications:

Neutralization, Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.