Please login to your online account to display your discounted pricing

anti-TFG, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224485

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224485

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Format Affinity Purified
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Primate
Cross Reactivity Rat
Concentration 0.5mg/mL
Primary or Secondary Primary
Test Specificity The USP13 amino acid sequence used as immunogen for the antibody is 100% homologous in human, chimpanzee, rhesus monkey, horse, mouse, cow and dog, 94% homologous in rat, 88% homologous in opossum and 82% homologous in chicken.
Gene ID (Entrez) 10342
Conjugate Unlabeled
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A portion of amino acids 350-400 of human TFG was used as the immunogen for this antibody.
Antigen TFG
Regulatory Status RUO
Quantity 0.1mg