Please login to your online account to display your discounted pricing

anti-TFEB, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10093447SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10093447T

  • / Each
 

Description & Specifications

Specifications

Applications Peptide ELISA
Applications Western Blotting
Quantity 0.02mg
Gene ID (Entrez) 7942
Host Species Goat
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Primary or Secondary Primary
Immunogen Peptide with sequence C-QKDLQKSRELENH
Antigen TFEB
Gene Accession No. P19484
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Test Specificity This antibody is expected not to cross-react to TFE3 and MITF.
Format Affinity Purified
Formulation Tris-saline (pH 7.3) with 0.5% BSA
Conjugate Unlabeled
Regulatory Status RUO