missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TET1, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human TET1 protein (between residues 170-800) [UniProt Q8NFU7]
Antigen TET1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP178965 Novus Biologicals Canada
NBP178965
0.1mL Each for $309.05
NBP178965SS Novus Biologicals Canada
NBP178965SS
0.025mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
TET1
Affinity Purified
RUO
Q8NFU7
PBS, 30% glycerol
A synthetic peptide made to an internal portion of the human TET1 protein (between residues 170-800) [UniProt Q8NFU7]
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin)
80312
1.07mg/mL