Please login to your online account to display your discounted pricing

anti-TET1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178966

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178966

  • / Each
 

Description & Specifications

Specifications

Antigen TET1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q8NFU7
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human TET1 protein (between residues 1700 to 1800) [UniProt Q8NFU7]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 80312
Target Species Murine