Please login to your online account to display your discounted pricing

anti-TET1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178965

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178965

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Gene Accession No. Q8NFU7
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 80312
Concentration 1.07mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Antigen TET1
Format Affinity Purified
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen A synthetic peptide made to an internal portion of the human TET1 protein (between residues 170-800) [UniProt Q8NFU7]
Quantity 0.1mL
Regulatory Status RUO