Please login to your online account to display your discounted pricing

anti-TERT, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11089471

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11089471

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Immunogen Synthetic peptide made to an internal portion of human Telomerase reverse transcriptase (within residues 900-950). [Swiss-Prot No. O14746]
Cross Reactivity Human
Cross Reactivity Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Host Species Rabbit
Antigen TERT
Test Specificity Human and mouse. Immunogen sequence has 84% homology to canine, rat and bovine proteins.
Gene ID (Entrez) 7015
Conjugate Unlabeled
Concentration 1mg/mL
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. O14746
Quantity 0.1mL
Regulatory Status RUO
Format Affinity Purified