Please login to your online account to display your discounted pricing

anti-TERT, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11089471SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11089471T

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Formulation Tris-citrate/phosphate, pH 7-8
Primary or Secondary Primary
Immunogen Synthetic peptide made to an internal portion of human Telomerase reverse transcriptase (within residues 900-950). [Swiss-Prot No. O14746]
Format Affinity Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Gene Accession No. O14746
Test Specificity Human and mouse. Immunogen sequence has 84% homology to canine, rat and bovine proteins.
Gene ID (Entrez) 7015
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Antigen TERT
Quantity 0.025mL