missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ anti-TER-119, FITC, Clone: TER-119,
Rat Monoclonal Antibody
$239.48 - $239.48
Specifications
Antigen | TER-119 |
---|---|
Clone | TER-119 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
MABF1434MI
|
MilliporeSigma™
MABF1434 |
25 μg |
Each for $239.48
|
|
|||||
Description
Specifically detects TER-119 Clone: TER-119 in Mouse samples, and it is validated for Flow Cytometry
The TER-119 antibody is named for the antigen to which it binds, a 52kDa surface protein that is associated with glycophorin-A. TER-119 is considered to be a lineage marker for later stages of erythroid cell development, as its expression begins at the pro-erythroblast stage. TER-119 antigen is not expressed at either BFU-E or CFU-E stages, i.e. prior to the pro-erythroblast stage.Specifications
TER-119 | |
Flow Cytometry | |
FITC | |
Rat | |
Inflammation, Immunology | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1%Â gelatin, pH 7.2. | |
Inpp5d, 7a33, Ship, Ship1 | |
Primary | |
2°C to 8°C for one year from date of shipment protected from light; Do not freeze |
TER-119 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
Q9ES52 | |
Proprietary immunogen | |
Purified by size-exclusion chromatography |
For Research Use Only