Please login to your online account to display your discounted pricing

anti-TER-119, Clone: TER-119, R&D Systems

$139.08 - $432.57

Specifications

Antigen TER-119
Clone TER-119
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MAB1125

 
r & d systems
RDSMAB1125
Unlabeled 500μg Each for $432.57

MLDP8

 
r & d systems
RDSMLDP8
Unlabeled 1 Vial Each for $139.08
Description & Specifications

Specifications

Antigen TER-119
Clone TER-119
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Regulatory Status RUO
Immunogen Mouse fetal liver cells.
Isotype IgG2b
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary

Applications:

Cell Depletion