Please login to your online account to display your discounted pricing

anti-TER-119, Clone: TER-119, R&D Systems


Manufacturer: r & d systems  RDSMLDP8

 View more versions of this product

Catalog No. MLDP8

  • / Each
 

Description & Specifications

Specifications

Antigen TER-119
Clone TER-119
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Immunogen Mouse fetal liver cells.
Isotype IgG2b
Quantity 1 Vial
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Murine