Please login to your online account to display your discounted pricing

anti-TDAG8/GPR65, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224487SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224487SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Immunogen A portion of amino acids 200-250 of human GPR65 was used as the immunogen for this antibody.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 8477
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human,monkey, cow, dog and chicken, 92% homologous in chimpanzee, 91% homologous in mouse and rat, 90% homologous in opossum and 88% homologous in pufferfish.
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Antigen TDAG8/GPR65
Format Affinity Purified