Please login to your online account to display your discounted pricing

anti-TDAG8/GPR65, Polyclonal, Novus Biologicals™

$109.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 200-250 of human GPR65 was used as the immunogen for this antibody.
Antigen TDAG8/GPR65
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224487SS

 
novus biologicals canada
NBP224487SS
Unlabeled 0.025mg Each for $109.00

NBP224487

 
novus biologicals canada
NBP224487
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 200-250 of human GPR65 was used as the immunogen for this antibody.
Antigen TDAG8/GPR65
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human,monkey, cow, dog and chicken, 92% homologous in chimpanzee, 91% homologous in mouse and rat, 90% homologous in opossum and 88% homologous in pufferfish.
Gene ID (Entrez) 8477
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL