Please login to your online account to display your discounted pricing

anti-TCF21, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11037257SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11037257T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. O43680
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Immunogen A synthetic peptide made to an internal region of human TCF21 protein (within residues 100-179). [Swiss-Prot No. O43680]
Primary or Secondary Primary
Gene ID (Entrez) 6943
Test Specificity Human, rat and bovine. Immunogen sequence has 90% homology to mouse and Xenopus.
Concentration 1mg/mL
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Rat
Antigen TCF21
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Host Species Rabbit
Quantity 0.025mL
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled