Please login to your online account to display your discounted pricing

anti-TCF21, Polyclonal, Novus Biologicals™

Manufacturer: novus biologicals canada  NB11037257

 View all options for this product


For Research Use Only

Catalog No. NB11037257


Description & Specifications


Antigen TCF21
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Rat
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. O43680
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of human TCF21 protein (within residues 100-179). [Swiss-Prot No. O43680]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6943
Test Specificity Human, rat and bovine. Immunogen sequence has 90% homology to mouse and Xenopus.