Please login to your online account to display your discounted pricing

anti-TCF21, Polyclonal, Novus Biologicals™

$139.00 - $139.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of human TCF21 protein (within residues 100-179). [Swiss-Prot No. O43680]
Antigen TCF21
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11037257T

 
novus biologicals canada
NB11037257SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of human TCF21 protein (within residues 100-179). [Swiss-Prot No. O43680]
Antigen TCF21
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human, rat and bovine. Immunogen sequence has 90% homology to mouse and Xenopus.
Gene Accession No. O43680
Gene ID (Entrez) 6943
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL