missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TC10 Polyclonal Antibody, Invitrogen™

TC10 Antibody, PA1-1061, from Invitrogen™. Species Reactivity: Human, Mouse; Applications: Immunoprecipitation, Western Blot.

Manufacturer:  Pierce Biotechnology Inc PA11061

Catalog No. PA1-1061


 

Specifications

Description

TC10 Antibody, PA1-1061, from Invitrogen. Species Reactivity: Human, Mouse; Applications: Immunoprecipitation, Western Blot.

TC10 has been classified as a member of the Rho family of GTPases and possesses characteristics most closely related to Cdc42. This family of proteins regulates a wide variety of cellular activity including cytoskeletal rearrangements, motility, proliferation and apoptosis. TC10 is predominately localized to the cell membrane and TC10 mRNA was most highly expressed in heart and skeletal muscle. Recent studies with TC10 mutations have demonstrated that TC10 is responsible for the activation of c-jun amino terminal kinase, the activation of serum response factor-dependent transcription, and the formation of long filopodia. In other studies, TC10 interacts with a similar subset of effectors as Cdc42, but does not react with WASP,mLK3, or ACK-1.
Host Species: Rabbit
Cross Reactivity: Human, Mouse
Isotype: IgG
Immunogen: Synthetic peptide corresponding D(136) P K T L A R L N D M K E K P I C(152) from human TC10.
Formulation: 100μg purified antibody (1mg/mL) in PBS with 1mg/mL BSA and 0.05% sodium azide
Synonyms: ARHQ, RASL7A, TC10, TC10A
Uniprot Accession: P17081, Q8R527
Entrez GeneID: 104215, 23433
Gene Symbol: RHOQ
PA1-1061 detects human and mouse TC10 protein in transfected cell samples and endogenous levels in tissue extracts. PA1-1061 is specific for isotype TC10a and demonstrates slight cross-reactivity with TC10b. PA1-1061 has successfully been used in immunoprecipitation and Western blot procedures. By Western blot, this antibody detects an ∽24 kDa protein representing TC10 in transfected COS cells and a non-specific, unidentified band at ∽50 kDa. The PA1-1061 immunogen is a synthetic peptide corresponding D(136) P K T L A R L N D M K E K P I C(152) from human TC10. This peptide (Cat. # PEP-219) is available for use in neutralization and control experiments.
Description & Specifications

Specifications

TC10
Unlabeled
Purified
P17081, Q8R527
RHOQ
Synthetic peptide corresponding D(136) P K T L A R L N D M K E K P I C(152) from human TC10.
100μg
Avoid Freeze/Thaw Cycles
Polyclonal
Human,Murine
Immunoprecipitation,Western Blot
Human,Mouse
100μg purified antibody (1mg/mL) in PBS with 1mg/mL BSA and 0.05% sodium azide
ARHQ, RASL7A, TC10, TC10A
Rabbit
IgG
Human, Mouse
Primary
104215, 23433
Promotions

Promotions

  • Purchase any three Invitrogen™ Antibodies and receive a fourth antibody at no additional cost. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.

    06/30/18