Please login to your online account to display your discounted pricing

anti-TBLR1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056501SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056501T

  • / Each
 

Description & Specifications

Specifications

Antigen TBLR1
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. Q9BZK7
Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide of human TBLR1
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 79718
Test Specificity A 61 kDa band and a 54 kDa band are observed. Cross reacts with Human. Predicted to react with Mouse and Rat based on BLAST results.
Target Species Human