Please login to your online account to display your discounted pricing

anti-TBLR1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056501

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056501

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.2% gelatin
Gene Accession No. Q9BZK7
Concentration 0.5mg/mL
Antigen TBLR1
Host Species Rabbit
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 79718
Test Specificity A 61 kDa band and a 54 kDa band are observed. Cross reacts with Human. Predicted to react with Mouse and Rat based on BLAST results.
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Applications Western Blotting
Immunogen This antibody was developed against a synthetic peptide of human TBLR1
Quantity 0.1mg