Please login to your online account to display your discounted pricing

anti-TBLR1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide of human TBLR1
Antigen TBLR1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056501T

 
novus biologicals canada
NB10056501SS
Unlabeled 0.025mg Each for $139.00

NB10056501

 
novus biologicals canada
NB10056501
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide of human TBLR1
Antigen TBLR1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity A 61 kDa band and a 54 kDa band are observed. Cross reacts with Human. Predicted to react with Mouse and Rat based on BLAST results.
Gene Accession No. Q9BZK7
Gene ID (Entrez) 79718
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL