Please login to your online account to display your discounted pricing

anti-TBKBP1, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human SINTBAD/TBKBP1 was used as the immunogen.
Antigen TBKBP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224465

 
novus biologicals canada
NBP224465
Unlabeled 0.1mg Each for $259.02

NBP224465SS

 
novus biologicals canada
NBP224465SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human SINTBAD/TBKBP1 was used as the immunogen.
Antigen TBKBP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and 94% homologous in chimpanzee.
Gene ID (Entrez) 9755
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL