Please login to your online account to display your discounted pricing

anti-TAX1BP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600975SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600975SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene Accession No. NP_001073333
Formulation PBS containing 0.05% BSA
Test Specificity There are multiple isoforms of TRAF6BP ranging from 590 amino acids to 858 amino acids. Researchers are encouraged to titer antibody for their specific applications. Cross-reacts with Human. Expected to cross-react with Mouse (95% identity with immunogen).
Concentration 0.5mg/mL
Cross Reactivity Human
Gene ID (Entrez) 8887
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Immunogen This antibody was developed against amino acids 718-735 (CSEQFPPDYDQQVFERHV) of human TRAF6BP (TAX1BP). The sequence used is 94% homologous in mouse.
Host Species Rabbit
Format Purified
Quantity 0.025mg
Antigen TAX1BP1
Regulatory Status RUO
Applications Western Blotting