Please login to your online account to display your discounted pricing

anti-Tankyrase 1, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a region of human Tankyrase within residues 250-350 [UniProt No. O95271]. The actual peptide sequence is proprietary.
Antigen Tankyrase 1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100186

 
novus biologicals canada
NB100186
Unlabeled 0.1mL Each for $309.05

NB100186SS

 
novus biologicals canada
NB100186SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a region of human Tankyrase within residues 250-350 [UniProt No. O95271]. The actual peptide sequence is proprietary.
Antigen Tankyrase 1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Gene Accession No. O95271
Gene ID (Entrez) 8658
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL