Please login to your online account to display your discounted pricing

anti-Tankyrase 1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100186SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100186SS

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-glycine, 150mM NaCl
Gene Accession No. O95271
Gene ID (Entrez) 8658
Concentration 1mg/mL
Antigen Tankyrase 1
Format Affinity Purified
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Immunogen A synthetic peptide made to a region of human Tankyrase within residues 250-350 [UniProt No. O95271]. The actual peptide sequence is proprietary.
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Cross Reactivity Human