Please login to your online account to display your discounted pricing

anti-Tankyrase 1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100186

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100186

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. O95271
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Gene ID (Entrez) 8658
Conjugate Unlabeled
Antigen Tankyrase 1
Format Affinity Purified
Immunogen A synthetic peptide made to a region of human Tankyrase within residues 250-350 [UniProt No. O95271]. The actual peptide sequence is proprietary.
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Host Species Rabbit
Quantity 0.1mL